Learn More
Wesbart™ Storage Bin
A useful way of keeping loose items safe within a freezer
1055.00 SEK - 1250.00 SEK
Products 4
Product Code Brand Height (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Height (Metric) Width (Metric) Price Quantity & Availability  
16246941
missing translation for 'viewDocuments' Promotion Details
Wesbart™
BIN923SS
280 mm 170 mm
1240.00 SEK
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 23-08-2024
to see stock.
Add to basket
15881118
missing translation for 'viewDocuments' Promotion Details
Wesbart™
BIN898
240 mm 190 mm
1250.00 SEK
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
15915590
missing translation for 'viewDocuments' Promotion Details
Wesbart™
BIN769
280 mm 140 mm
1060.00 SEK
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
15925590
missing translation for 'viewDocuments' Promotion Details
Wesbart™
BIN827
280 mm 280 mm
1055.00 SEK
Each
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • Storage bins are manufactured from anodised aluminum or stainless steel
  • All storage bins have either a fold down handle or hand grip
  • Bins can be designed to have a handle on each end for transport if required
  • All storage bins are fitted with a label holder so the user can easily identify which bin they require
  • Videos