Learn More
Gosselin™ Straight Assembled Sterile Container with Red screw Cap
Polypropylene assembled straight container with screw cap
3075.00 SEK - 7680.00 SEK
Specifications
Material | Polypropylene |
---|---|
Sterility | Sterile |
Closure Color | Red |
Closure Type | Screw Cap with Inner Lip |
Color | Clear |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15408794
|
Gosselin™
BTP30C-002_CE_IVD |
40 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15418794
|
Gosselin™
BTP35C-002_CE_IVD |
60 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15428794
|
Gosselin™
BTP53C-002_CE_IVD |
180 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Polypropylene | |
Red | |
Clear |
Sterile | |
Screw Cap with Inner Lip |