Learn More
Abcam Streptavidin Coated Plate
Product Code. 32115096 Shop All Abcam Products
Change view
Click to view available options
Quantity:
5 x 96 wells
Unit Size:
Pack of 96
Product Code. Quantity Unit Size
32115096 5 x 96 wells Pack of 96
1 options
Product Code. 32115096 Supplier Abcam Supplier No. ab3234955x96Wells
 Restricted Item
Beräknad leverans: 01-12-2026
Plate Characteristics:
Biotin Binding Protein, Plate Type, Coat Volume, Block Volume, Approximate Binding Capacity, Steptavidin
Plate Characteristics:
Biotin Binding Protein, Plate Type, Coat Volume, Block Volume, Approximate Binding Capacity Steptavidin, 96-well Strip, 100 μL/well, 300 μL/well, ~10 pmol Biotin/well
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 5 x 96 wells
Regulatory Status RUO