missing translation for 'onlineSavingsMsg'
Learn More

STT3A Mouse anti-Human, Mouse, Clone: 1E2B12, Proteintech

Mouse Monoclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen STT3A
Clone 1E2B12
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16888585
View Documents
Proteintech
66581-1-IG-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16878585
View Documents
Proteintech
66581-1-IG-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes the basic transcription factor 3. This protein forms a stable complex with RNA polymerase IIB and is required for transcriptional initiation. Alternative splicing results in multiple transcript variants encoding different isoforms. This gene has multiple pseudogenes.
Specifications

Specifications

STT3A
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
B5, Integral membrane protein 1, ITM1, STT3 A, STT3A, TMC, Transmembrane protein TMC
STT3A
IgG1
Protein G
Antibody
1E2B12
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human, Mouse
P46977, P46978
16430, 3703
STT3A Fusion Protein Ag27072
Primary
-20°C
STT3A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
STT3A Mouse anti-Human, Mouse, Clone: 1E2B12, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.