missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUCLA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12627-1-AP-150UL
This item is not returnable.
View return policy
Description
SUCLA2 (succinate-CoA ligase, ADP-forming, beta subunit), also known as A-BETA, SCS-betaA or renal carcinoma antigen NY-REN-39, is a 463 amino acid mitochondrial matrix enzyme that belongs to the succinate/malate CoA ligase beta subunit family. Widely expressed, SUCLA2 dimerizes with the SCS alpha subunit to form SCS-A, an essential component of the tricarboxylic acid cycle. Defects in SUCLA2 may be involved in a group of autosomal recessive disorders known as Mitochondrial DNA depletion syndromes (MDSs) that are characterized by a decrease in mitochondrial DNA copy numbers in affected tissues.Specifications
SUCLA2 | |
Polyclonal | |
Unconjugated | |
SUCLA2 | |
A BETA, SCS betaA, SUCLA2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20916, 361071, 8803 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9P2R7, Q9Z2I9 | |
Sucla2 | |
SUCLA2 Fusion Protein Ag3319 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SUCLA2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction