Learn More
SUFU Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26759-1-AP-20UL
 View more versions of this product
Product Code. 16885125
  • 1645.54 SEK / 20µL
Description
This gene is a member of the H/ACA snoRNPs gene family. snoRNPs are involved in various aspects of rRNA processing and modification and have been classified into two families: C/D and H/ACA. The H/ACA snoRNPs also include the NOLA1, 2 and 3 proteins. The protein encoded by this gene and the three NOLA proteins localize to the dense fibrillar components of nucleoli and to coiled bodies in the nucleus. Both 18S rRNA production and rRNA pseudouridylation are impaired if any one of the four proteins is depleted. These four H/ACA snoRNP proteins are also components of the telomerase complex. The protein encoded by this gene is related to the Saccharomyces cerevisiae Cbf5p and Drosophila melanogaster Nop60B proteins. The gene lies in a tail-to-tail orientation with the palmitoylated erythrocyte membrane protein gene and is transcribed in a telomere to centromere direction. Both nucleotide substitutions and single trinucleotide repeat polymorphisms have been found in this gene. Mutations in this gene cause X-linked dyskeratosis congenita, a disease resulting in reticulate skin pigmentation, mucosal leukoplakia, nail dystrophy, and progressive bone marrow failure in most cases. Mutations in this gene also cause Hoyeraal-Hreidarsson syndrome, which is a more severe form of dyskeratosis congenita. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
SUFU
Polyclonal
Unconjugated
SUFU
PRO1280, SUFU, SUFUH, SUFUXL, Suppressor of fused homolog
Rabbit
Antigen Affinity Chromatography
RUO
24069, 361769, 51684
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunoprecipitation
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UMX1, Q9Z0P7
Sufu
SUFU Fusion Protein Ag25200
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SUFU Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated