Learn More
SULT1E1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12522-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody detected a major band around 35 kDa in lysates of mouse kidney and human liver.
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a protein that transfers a sulfo moiety to and from estrone, which may control levels of estrogen receptors.Specifications
SULT1E1 | |
Polyclonal | |
Unconjugated | |
SULT1E1 | |
EST, EST 1, Estrogen sulfotransferase, ST1E1, STE, Sulfotransferase 1E1, SULT1E1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20860, 360268, 6783 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.38 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P49888, P49891, P52844 | |
Sult1e1 | |
SULT1E1 Fusion Protein Ag3216 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Product Title
SULT1E1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction