missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT4A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SULT4A1 |
---|---|
Concentration | 0.39 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890354
|
Proteintech
12578-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880354
|
Proteintech
12578-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure(number and length of exons) is similar among family members. SULT4A1 isoforms a and b are selectively expressed in brain tissue. Isoform a is the predomit of the two isoforms of this protein. Isoform b includes an alternate segment, compared to isoform a, that causes a frameshift. The resulting protein (isoform b) has a distinct and shorter C-terminus when compared to isoform a.Specifications
SULT4A1 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
P63046, P63047, Q9BR01 | |
25830, 29859, 58953 | |
SULT4A1 Fusion Protein Ag3271 | |
Primary | |
-20°C | |
SULT4A1 |
0.39 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BR STL 1, BRSTL1, hBR STL, hBR STL 1, NST, ST4A1, Sulfotransferase 4A1, SULT4A1, SULTX3 | |
Sult4a1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SULT4A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech