Learn More
SUMF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25015-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes an enzyme that catalyzes the hydrolysis of sulfate esters by oxidizing a cysteine residue in the substrate sulfatase to an active site 3-oxoalanine residue, which is also known as C-alpha-formylglycine. Mutations in this gene cause multiple sulfatase deficiency, a lysosomal storage disorder. Alternative splicing results in multiple transcript variants.Specifications
SUMF1 | |
Polyclonal | |
Unconjugated | |
SUMF1 | |
AAPA3037, FGE, sulfatase modifying factor 1, SUMF1, UNQ3037/PRO9852 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
285362, 58911 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8NBK3, Q8R0F3 | |
SUMF1 | |
SUMF1 Fusion Protein Ag19998 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
SUMF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction