Learn More
SUN1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24568-1-AP-20UL
 View more versions of this product
Product Code. 16883045
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
This gene is a member of the unc-84 homolog family and encodes a nuclear envelope protein with an Unc84 (SUN) domain. The protein is involved in nuclear anchorage and migration. Several alternatively spliced transcript variants of this gene have been described; however, the full-length nature of some of these variants has not been determined.
Specifications
Specifications
SUN1
Polyclonal
Unconjugated
SUN1
KIAA0810, Protein unc 84 homolog A, Sad1/unc 84 protein like 1, SUN1, unc 84 homolog A (C. elegans), UNC84A
Rabbit
Antigen Affinity Chromatography
RUO
23353
-20°C
Liquid
Western Blot, Immunoprecipitation
0.22 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O94901
SUN1
SUN1 Fusion Protein Ag21117
20 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SUN1 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated