Learn More
Memmert™ Support Baskets for Water Bath
Use these baskets with Memmert ovens. Memmert™ Support Baskets for Water Bath feature stainless steel perforated mounting shelves with grid perforations to take clips for conical flasks.
Brand: Memmert™ B04397
This item is not returnable.
View return policy
Description
- Stainless steel support baskets
- For Memmert Water Baths
Specifications
Support Basket | |
Stainless Steel perforated mounting shelf with grid perforations |
Product Content Correction
Product Title
Memmert™ Support Baskets for Water Bath > For use with 29L and 45L baths
Spot an opportunity for improvement?Share a Content Correction