Learn More
SVL™ Cone/Screw Thread Adapter
Facilitates connection between SVL™ and Quickfit™ glassware.
3375.00 SEK - 4915.00 SEK
Product Code | Brand | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Price | Quantity & Availability | ||||||
15577740
|
SVL
670-70 |
4915.00 SEK
Pack of 10 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||||
15587740
|
SVL
670-77 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||||
15597740
|
SVL
670-83 |
Cone size: 29/32; Joint size: 30 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Features an SVL™ screwthread and Quickfit™ ground glass socket
- Thread accepts SVL™ screw caps and couplings. Cone fits Quickfit™ sockets
- Manufactured from temperature and chemical resistant borosilicate glass