Learn More
Hettich™ Swing-out Rotor for ROTOFIX 32 A and Universal 320/320 R Centrifuge, 12-place
Designed for an extended range of applications
Brand:  Hettich™ 1628
Product Code. 10664673
  • 2845.00 SEK / Each
Description
  • Swing-out Rotor, 12-place
  • For ROTOFIX 32 A and Universal 320/320 R Centrifuge
  • 55° (with carriers 1127-A)
  • 60° (with carriers 1122)
  • 80° (with carriers 1621)
Specifications
Specifications
Swing-out Rotor
5,000 rpm
125 mm
55/60/80°
4,193 x g
150 mm
ROTOFIX 32 A and Universal 320/320 R Centrifuge
12
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hettich™ Swing-out Rotor for ROTOFIX 32 A and Universal 320/320 R Centrifuge, 12-place > For ROTOFIX 32 A and Universal 320/320 R Centrifuge