missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptogyrin-4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | Synaptogyrin-4 |
---|---|
Concentration | 0.24 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16891345
|
Proteintech
21751-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16881345
|
Proteintech
21751-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Synaptogyrins comprise a family of tyrosine-phosphorylated membrane proteins with two neuronal (SYNGR1 and SYNGR3) and one ubiquitous (SYNGR2) members. SYNGR1 and -3 are synaptic vesicle proteins, residing in some cases on the same synaptic vesicle, and are thought to be involved in the regulation of neurotransmitter release. SYNGR2, by contrast, is absent from synaptic vesicles. SYNGR3 is predomitly expressed in brain and placenta. The role and localization of a fourth synaptogyrin, SYNGR4, is unclear.Specifications
Synaptogyrin-4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
O95473, Q9Z1L2 | |
23546, 292916, 58867 | |
Synaptogyrin-4 Fusion Protein Ag15865 | |
Primary | |
-20°C | |
SYNGR4 |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
synaptogyrin 4, Synaptogyrin-4, SYNGR4 | |
Syngr4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Synaptogyrin-4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech