Learn More
Epredia™ Lab Vision™ Synaptophysin Ab-2, Mouse Monoclonal Antibody
Provide accurate, reproducible results with the Epredia™ Synaptophysin Ab-2, Mouse Monoclonal Antibody.
Brand: Epredia™ MS-1150-R7
Additional Details : Weight : 0.01200kg
Description
Host Species: Mouse
Clone: SYP02
Isotype: IgG1
Species Reactivity: Human and Rat. Others not known.
Epitope: Near C-terminus
Immunogen: A synthetic peptide encoding a region near the C-terminal end of the synaptophysin protein
Molecular Weight: 38kDa
Positive Control: Pancreas, rheochromocytoma or thyroid
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Synaptophysin Ab-2 | |
Monoclonal | |
Unconjugated | |
A synthetic peptide encoding a region near the C-terminal end of the synaptophysin protein | |
RUO | |
Primary | |
Prediluted |
Immunohistochemistry (Paraffin) | |
SYP02 | |
Mouse | |
7 mL | |
Neuroscience | |
Human, Rat | |
IgG1 |