Learn More
Epredia™ Lab Vision™ Synaptophysin Ab-2, Mouse Monoclonal Antibody
Provide accurate, reproducible results with the Epredia™ Synaptophysin Ab-2, Mouse Monoclonal Antibody.
1203.60 SEK - 4391.10 SEK
Specifications
Antigen | Synaptophysin Ab-2 |
---|---|
Clone | SYP02 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12649116
|
Epredia™
MS-1150-S0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12659116
|
Epredia™
MS-1150-S1 |
Concentrated | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12639116
|
Epredia™
MS-1150-S |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12629116
|
Epredia™
MS-1150-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Host Species: Mouse
Clone: SYP02
Isotype: IgG1
Species Reactivity: Human and Rat. Others not known.
Epitope: Near C-terminus
Immunogen: A synthetic peptide encoding a region near the C-terminal end of the synaptophysin protein
Molecular Weight: 38kDa
Positive Control: Pancreas, rheochromocytoma or thyroid
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Synaptophysin Ab-2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human, Rat | |
IgG1 |
SYP02 | |
Monoclonal | |
Mouse | |
Neuroscience | |
A synthetic peptide encoding a region near the C-terminal end of the synaptophysin protein | |
Primary |