missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptotagmin-4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12642-1-AP-150UL
This item is not returnable.
View return policy
Description
May be involved in Ca2+-dependent exocytosis of secretory vesicles through Ca2+ and phospholipid binding to the C2 domain or may serve as Ca2+ sensors in the process of vesicular trafficking and exocytosis. Plays a role in dendrite formation by melanocytes.Specifications
Synaptotagmin-4 | |
Polyclonal | |
Unconjugated | |
SYT4 | |
HsT1192, KIAA1342, Synaptotagmin 4, synaptotagmin IV, Synaptotagmin-4, SYT4, SytIV | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20983, 64440, 6860 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P40749, P50232, Q9H2B2 | |
Syt4 | |
Synaptotagmin-4 Fusion Protein Ag3338 | |
150 ÎĽL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Synaptotagmin-4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 ÎĽL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction