Learn More
Syntaxin 16 Mouse anti-Human, CoraLite 488, Clone: 4A9D8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66775-100UL
Product Code. 16810846
  • 5106.85 SEK / 100µL
This item is not returnable. View return policy
Description
Description
The Syntaxin 16 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Syntaxin 16. This antibody recognizes human antigen. The Syntaxin 16 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
Syntaxin 16
Monoclonal
CoraLite 488
100 μL
8675
IgG1
Immunofluorescence
4A9D8
Mouse
Primary
Human
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Syntaxin 16 Mouse anti-Human, CoraLite 488, Clone: 4A9D8, Proteintech > 100 μL