Learn More
Novus Biologicals™ Syntaxin Binding Protein 4 293T Cell Transient Overexpression Lysate
Brand:  Novus Biologicals™ H00252983-T01
Product Code. 18486419
  • 4495.00 SEK / 0.10mL
This item is not returnable. View return policy
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.1 mL
Endocrinology, Signal Transduction
FLJ16496, MGC50337, ST x 4-interacting protein, SynipMGC149829, syntaxin 4 interacting protein, Syntaxin 4-interacting protein, syntaxin binding protein 4, syntaxin-binding protein 4
Store at -80°C. Avoid freeze/thaw cycles.
Syntaxin Binding Protein 4
STXBP4
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Syntaxin Binding Protein 4 293T Cell Transient Overexpression Lysate > 0.1 mL