Learn More
T Cell Receptor Gamma/Delta Hamster, anti-Mouse, Biotin, Clone: GL3, STEMCELL Technologies
Hamster Monoclonal Antibody
Brand:  Stemcell Technologies 60104BT
Encompass_Preferred
 View more versions of this product
Product Code. 17199021
  • 7303.50 SEK / 500µg
Description
  • The GL3 antibody reacts with the δ chain of the murine T cell receptor γ/δ (TCRγ/δ or TCR gamma/delta), a subtype of the TCR involved in the recognition of both peptide and lipid antigens
  • TCRγ/δ comprises a heterodimer (∼ 80 kDa in humans) of disulfide-linked γ and δ subunits that associates with CD3 on the cell surface
  • It is a member of the immunoglobulin superfamily
  • TCRγ/δ is expressed on a subpopulation of T cells in the circulation but may be found on up to 50% of the T cells in epithelial cell-rich tissues
  • These cells have roles in oral and tumor-associated tolerance as well as autoimmune disease and have been described as a link between the adaptive and innate immune responses
  • Once activated, they secrete effector cytokines in a subtype- and context-specific manner
  • Most γ/δ T cells are CD4-/CD8-, though some express CD8
  • The GL3 antibody recognizes an epitope in the constant region of the δ chain and can reportedly activate TCRγ/δ+ cells
Specifications
Specifications
T Cell Receptor Gamma/Delta
Monoclonal
Biotin
500 μg
Mouse
T Cells, T Cells, Other Subsets
Immunohistochemistry, Flow Cytometry, Immunofluorescence, Functional Assay, Blocking Assay
GL3
Hamster
Primary
IgG2 κ
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
T Cell Receptor Gamma/Delta Hamster, anti-Mouse, Biotin, Clone: GL3, STEMCELL Technologies