missing translation for 'onlineSavingsMsg'
Learn More

TAF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 20260-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16889784

  • 4721.67 SEK / 150µL
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The antibody is specific to TAF1.

3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.
Specifications

Specifications

TAF1
Polyclonal
Unconjugated
TAF1
BA2R, CCG1, CCGS, Cell cycle gene 1 protein, DYT3, KAT4, N TAF1, NSCL2, OF, P250, TAF(II)250, TAF1, TAF2A, TAFII 250, TAFII250, TBP associated factor 250 kDa
Rabbit
Antigen Affinity Chromatography
RUO
270627, 6872
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P21675, Q80UV9
TAF1
Peptide
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers