missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF9B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10921-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine. The encoded protein is involved in myelopoiesis, and defects in this gene have been associated with several myeloproliferative disorders. Two variants encoding different isoforms have been found for this gene.Specifications
TAF9B | |
Polyclonal | |
Unconjugated | |
TAF9B | |
DN 7, DN7, TAF9B, TAF9L, TAFII31L, TFIID 31 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
171152, 407786, 51616 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q62880, Q6NZA9, Q9HBM6 | |
Taf9b | |
TAF9B Fusion Protein Ag1362 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |