missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF9B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | TAF9B |
---|---|
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827633
|
Proteintech
10921-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16817633
|
Proteintech
10921-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a methylcytosine dioxygenase that catalyzes the conversion of methylcytosine to 5-hydroxymethylcytosine. The encoded protein is involved in myelopoiesis, and defects in this gene have been associated with several myeloproliferative disorders. Two variants encoding different isoforms have been found for this gene.Specifications
TAF9B | |
Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q62880, Q6NZA9, Q9HBM6 | |
171152, 407786, 51616 | |
TAF9B Fusion Protein Ag1362 | |
Primary | |
-20°C | |
TAF9B |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DN 7, DN7, TAF9B, TAF9L, TAFII31L, TFIID 31 | |
Taf9b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |