missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TAK 21d
Potent FAAH inhibitor
1970.00 SEK - 7470.00 SEK
Specifications
Chemical Name or Material | 4-[4-(3,4-Difluorophenyl)-2-pyrimidinyl]-N-3-pyridazinyl-1-piperazinecarboxamide |
---|---|
CAS | 1143578-94-2 |
Target | Fatty Acid Amide Hydrolase (FAAH) Inhibitors |
Molecular Formula | C19H17F2N7O |
Formula Weight | 397.38 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18768589
|
Tocris Bioscience™
5209/10 |
10 mg |
1970.00 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18776138
|
Tocris Bioscience™
5209/50 |
50 mg |
7470.00 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent FAAH inhibitor; (IC50 values are 0.28 and 0.72 nM, at rat and human FAAH respectively). Displays analgesic effects in vivo models of neuropathic and inflammatory pain. Brain penetrant.Specifications
4-[4-(3,4-Difluorophenyl)-2-pyrimidinyl]-N-3-pyridazinyl-1-piperazinecarboxamide | |
Fatty Acid Amide Hydrolase (FAAH) Inhibitors | |
397.38 |
1143578-94-2 | |
C19H17F2N7O | |
>98% |
Safety and Handling
Recommended Storage : Store at -20°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tocris Bioscience™ TAK 21d