Läs mer
Tocris Bioscience™ TAK 960 hydrochloride
GREENER_CHOICE
Potent and selective PLK1 inhibitor
Brand:  Tocris Bioscience™ 5403/50
Encompass
Produktkod. 15756796
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter »
Beskrivning
Beskrivning
TAK 960 hydrochloride is a potent PLK1 inhibitor (IC50 values are 0.8, 16.9 and 50.2 nM for PLK1, PLK2 and PLK3 respectively). Exhibits >20-fold selectivity for PLK1 over FAK, MLCK and FES, and has minimal activity against a panel of 282 other kinases. Inhibits proliferation of a range of cancer cell lines in vitro and inhibits tumor growth of multiple human cancer cell xenografts in vivo. Orally bioavailable.
Specifikationer
Specifikationer
>98%
TAK 960 hydrochloride
TAK 960 hydrochloride
2108449-45-0
598.06
50 mg
Produktförslag
Produktförslag
Filmer
SDS
Dokument
Dokument
Certifikat
Kampanjer
Kampanjer