missing translation for 'onlineSavingsMsg'
Learn More

TBC1D5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17078-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16836914

  • 1656.73 SEK / 20µL
Estimated Shipment: 14-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TBC1 domain family member 5 (TBC1D5) may act as a GTPase-activating protein for Rab family protein(s).
Specifications

Specifications

TBC1D5
Polyclonal
Unconjugated
TBC1D5
KIAA0210, TBC1 domain family member 5, TBC1 domain family, member 5, TBC1D5
Rabbit
Antigen Affinity Chromatography
RUO
501088, 72238, 9779
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q80XQ2, Q92609
Tbc1d5
TBC1D5 Fusion Protein Ag10991
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers