missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | TBC1D5 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16836914
|
Proteintech
17078-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16826914
|
Proteintech
17078-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TBC1 domain family member 5 (TBC1D5) may act as a GTPase-activating protein for Rab family protein(s).Specifications
TBC1D5 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q80XQ2, Q92609 | |
501088, 72238, 9779 | |
TBC1D5 Fusion Protein Ag10991 | |
Primary | |
-20°C | |
TBC1D5 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA0210, TBC1 domain family member 5, TBC1 domain family, member 5, TBC1D5 | |
Tbc1d5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |