Learn More
TBL1X Mouse anti-Human, Unconjugated, Clone: 1A9A1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66955-1-IG-150UL
 View more versions of this product
Product Code. 16879275
  • 4652.90 SEK / 150µL
Description
The TBL1X antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TBL1X. This antibody recognizes human, mouse, rat antigen. The TBL1X antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
TBL1X
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1A9A1
Unconjugated
150 μL
6907
IgG2a
Certificates
Promotions
Promotions
Product Content Correction
Product Title
TBL1X Mouse anti-Human, Unconjugated, Clone: 1A9A1, Proteintech > 150 μL