missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22246-1-AP-150UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody raised against part of N-terminal chain of human TBP, and specifically recognize a ∽46 kDa band.
RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA, RAC1, and CDC42.Specifications
TBP | |
Polyclonal | |
Unconjugated | |
TBP | |
GTF2D, GTF2D1, SCA17, TATA binding factor, TATA box binding protein, TATA box factor, TATA sequence binding protein, TBP, TF2D, TFIID | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21374, 6908 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20226, P29037 | |
TBP | |
TBP Fusion Protein Ag17740 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TBP Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction