missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBRG4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TBRG4 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895814
|
Proteintech
16245-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16885814
|
Proteintech
16245-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TBRG4 (transforming growth factor beta regulator 4), also known as CPR2 (cell cycle progression restoration protein 2) or FAST 4 (FAST kinase domain-containing protein 4), is a 631 amino acid protein that contains one RAP domain and belongs to the FAST kinase family. TBRG4 is ubiquitously expressed and may have a role in cell cycle progression. Existing as two alternatively spliced isoforms, the gene encoding TBRG4 maps to human chromosome 7p13.Specifications
TBRG4 | |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q5M9G9, Q91YM4, Q969Z0 | |
21379, 360977, 9238 | |
TBRG4 Fusion Protein Ag9176 | |
Primary | |
-20°C | |
TBRG4 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CPR2, FASTKD4, KIAA0948, Protein TBRG4, TBRG4 | |
Tbrg4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TBRG4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech