Learn More
TDP-43 (C-terminal) Mouse anti-Human, Unconjugated, Clone: 1G5F6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67345-1-IG-20UL
 View more versions of this product
Product Code. 16889925
  • 1645.54 SEK / 20µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
The TDP-43 (C-terminal) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human TDP-43 (C-terminal). This antibody recognizes human, mouse, rat antigen. The TDP-43 (C-terminal) antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
Specifications
TDP-43 (C-terminal)
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry
1G5F6
Unconjugated
20 ÎĽL
23435
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TDP-43 (C-terminal) Mouse anti-Human, Unconjugated, Clone: 1G5F6, Proteintech > 20 ÎĽL