Learn More
TDP-43 Rabbit anti-Human, Unconjugated, Clone: 11N20, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80001-1-RR-100UL
Additional Details : Weight : 0.00010kg
Description
The TDP-43 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human TDP-43. This antibody recognizes human, mouse, rat antigen. The TDP-43 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
TDP-43 | |
Monoclonal | |
250 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Flow Cytometry | |
11N20 | |
Unconjugated | |
100 μL | |
23435 | |
IgG |