missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDP-43 Rabbit anti-Human, Unconjugated, Clone: 11N20, Proteintech
Rabbit Recombinant Antibody
1763.23 SEK - 5195.02 SEK
Specifications
Antigen | TDP-43 |
---|---|
Clone | 11N20 |
Concentration | 250 μg/mL |
Applications | Western Blot, Immunohistochemistry, Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840426
|
Proteintech
80001-1-RR-20UL |
20 μL |
1763.23 SEK
20µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16830426
|
Proteintech
80001-1-RR-100UL |
100 μL |
5195.02 SEK
100µL |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descrizione
The TDP-43 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human TDP-43. This antibody recognizes human, mouse, rat antigen. The TDP-43 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifica
TDP-43 | |
250 μg/mL | |
Monoclonal | |
Rabbit | |
23435 | |
Primary |
11N20 | |
Western Blot, Immunohistochemistry, Flow Cytometry | |
Unconjugated | |
Human, Rat, Mouse | |
IgG |