Learn More
TDP-43 Rabbit anti-Human, Unconjugated, Clone: 11N20, Proteintech
Rabbit Recombinant Antibody
Brand:  Proteintech 80001-1-RR-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840426
  • 1763.23 SEK / 20µL
Estimated Shipment: 30-07-2024
to see stock.
Description
Description
The TDP-43 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human TDP-43. This antibody recognizes human, mouse, rat antigen. The TDP-43 antibody has been validated for the following applications: FC, IHC, WB, ELISA analysis.
Specifications
Specifications
TDP-43
Monoclonal
250 μg/mL
Rabbit
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Flow Cytometry
11N20
Unconjugated
20 μL
23435