Läs mer
Tegafur-13C,15N2, TRC
Produktkod. 30416205
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30416205 10 mg
30421364 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30416205 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT01390310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1189456-27-6
Molekylformel 13C C7 H9 F 15N2 O3
Molekylvikt (g/mol) 203.15
Synonym Tegafur 13C,15N2
IUPAC-namn 5-fluoro-1-(oxolan-2-yl)(2^{13}C,1,3-^{15}N_{2})pyrimidine-2,4-dione
LEDER FC1=C[15N](C2CCCO2)[13C](=O)[15NH]C1=O
Specifikationer
Kemiskt namn eller material Tegafur-13C,15N2
Kvantitet 10 mg
InChI formel InChI=1S/C8H9FN2O3/c9-5-4-11(6-2-1-3-14-6)8(13)10-7(5)12/h4,6H,1-3H2,(H,10,12,13)/i8+1,10+1,11+1
Formel vikt 203.0571