missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ Analog Output

For Use With: All MK Series models

Brand:  BINDER™ 8012-1084

12494.75 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15984252

  • 13745.00 SEK / Each
Only null left
Estimated Shipment: 12-11-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Output not adjustable
  • 4 to 20 mA
Specifications

Specifications

Analog output 4-20 mA - for temperature values (output not adjustable)
Plastic
Temperature
BINDER Chambers Series MK
Analog Output
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BINDER™ Analog Output > For Use With: All MK Series models

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.