Learn More
BINDER™ Temperature Analog Output
For Use With: BINDER units BD 56, BD 115, BD 260, BD 720
Brand: BINDER™ 8012-0941
12494.75 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Output not adjustable
- 4 to 20 mA
Specifications
Analog output 4-20 mA - for temperature values (output not adjustable) | |
Plastic | |
Temperature |
BINDER Chambers BD 56, BD 115, BD 260, BD 720 | |
Analog Output |
Product Content Correction
Product Title
BINDER™ Temperature Analog Output > For Use With: BD 56, BD 720, BD 115, BD 260, BF 115, ED 56, ED 115, ED 260, FD 56, FD 115, FD 260, FED 260
Spot an opportunity for improvement?Share a Content Correction