Learn More
Fisher Chemical 500 ML Ten elements solution - B, Ge, Mo, P Sb Si Sn, Te, Ti, Zr forICP 10mg/L of each 2% HNO3 0.1%
Brand:  Fisher Chemical CM1738
500 ML Ten elements solution - B, Ge, Mo, P Sb Si Sn, Te, Ti, Zr forICP 10mg/L of each 2% HNO3 0.1%
Additional Details : Weight : 0.50000kg
Product Code. 12914345