missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Propionate EP Impurity B, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Testosterone Propionate EP Impurity B
Specifications
Specifications
Chemical Name or Material | 3-Oxoandrost-4-en-17beta-yl 2-Methyl-propanoate (Testosterone Isobutyrate) |
CAS | 1169-49-9 |
Analyte or Component Names | 3-Oxoandrost-4-en-17beta-yl 2-Methyl-propanoate (Testosterone Isobutyrate) |
Shipping Condition | Room Temperature |
Molecular Formula | C23 H34 O3 |
Certifications/Compliance | ISO 9001 |
Synonym | 3-Oxoandrost-4-en-17β-yl 2-Methyl-propanoate (Testosterone Isobutyrate), Testosterone Isocaproate Imp. D (EP), Testosterone Propionate Imp. B (EP), 3-Oxoandrost-4-en-17beta-yl 2-Methylpropanoate, Testosterone isobutyrate, Androst-4-en-3-one, 17-(2-methyl-1-oxopropoxy)-, (17beta)-, Testosterone, isobutyrate (6CI,7CI,8CI), (17beta)-17-(2-Methyl-1-oxopropoxy)androst-4-en-3-one, Isobutyric acid, ester with testosterone (8CI), 17beta-(Isobutyroxy)androst-4-en-3-one, 3-Oxoandrost-4-en-17beta-yl isobutyrate, Agovirin-Depot, Perandren M, Testocryst, Testosterone 17-isobutyrate, Testosterone 2-methylpropanoate, Testosterone 2-methylpropionate, Testosterone Isocaproate Impurity D, Testosterone Propionate Impurity B |
InChI Formula | InChI=1S/C23H34O3/c1-14(2)21(25)26-20-8-7-18-17-6-5-15-13-16(24)9-11-22(15,3)19(17)10-12-23(18,20)4/h13-14,17-20H,5-12H2,1-4H3/t17-,18-,19-,20-,22-,23-/m0/s1 |
SMILES | CC(C)C(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
IUPAC Name | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 2-methylpropanoate |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Testosterone Propionate EP Impurity B, Mikromol™ >
Spot an opportunity for improvement?Share a Content Correction