missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA572805
This item is not returnable.
View return policy
Description
Dioxygenase that specifically binds methylcytosine (5mC), a minor base in mammalian DNA found in repetitive DNA elements that is crucial for retrotransposon silencing and mammalian development. Catalyzes the conversion of methylcytosine (5mC) to 5-hydroxymethylcytosine (hmC). The clear function of 5-hydroxymethylcytosine (hmC) is still unclear but it may influence chromatin structure and recruit specific factors or may constitute an intermediate component in cytosine demethylation. 5-hydroxymethylcytosine (hmC) is present in ES cells and is enriched in the brain, especially in Purkinje neurons. May play a role in the fetal development of heart, lung and brain.
Specifications
TET1 | |
Polyclonal | |
Unconjugated | |
TET1 | |
2510010B09Rik; AA517754; bA119F7.1; BB001228; CXXC finger 6; CXXC zinc finger 6; CXXC6; CXXC-type zinc finger protein 6; D10Ertd17e; KIAA1676; LCX; leukemia-associated protein with a CXXC domain; Methylcytosine dioxygenase TET1; mKIAA1676; MLL-TET1; RP11-119F7.1; Ten-eleven translocation 1 gene protein; ten-eleven translocation 1 gene protein homolog; ten-eleven translocation-1; tet methylcytosine dioxygenase 1; tet oncogene 1; Tet1; TET1 splice variant VP_DE4; TET1 splice variant VP_DE456; TET1-MLL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
309902, 52463, 80312 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q3URK3, Q8NFU7 | |
TET1 | |
TET1 antibody for an 18 amino acid peptide near the carboxy terminus of human TET1. The immunogen is within amino acids 2030 - 2080 of TET1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TET1 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction