missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TEX38 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TEX38 |
---|---|
Concentration | 0.42 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16882905
|
Proteintech
24429-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16872905
|
Proteintech
24429-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody detects 25-28 kDa with mouse testis and MDA-MB-453s cell lysate.
TEX38 is a protein coding gene.Specifications
TEX38 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
A2A8T7, Q6PEX7 | |
374973, 75173 | |
TEX38 Fusion Protein Ag19833 | |
Primary | |
-20°C | |
TEX38 |
0.42 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ATPAF1-AS1, C1orf223, TEX38 | |
TEX38 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TEX38 Rabbit anti-Human, Mouse, Polyclonal, Proteintech