Learn More
Molecular Probes™ Texas Red™ C2-Dichlorotriazine
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
Texas Red C2-dichlorotriazine is a reactive dye with absorption/emission maxima of ∼588/601 nm. Dichlorotriazines readily modify amines in proteins, and are among the few reactive groups that are reported to react directly with polysaccharides and other alcohols in aqueous solution, provided that the pH is >9 and that other nucleophiles are absent.
Specifications
Specifications
Quantity | 5 mg |
Product Type | C2-Dichlorotriazine |
Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
Chemical Reactivity | Alcohol, Amine |
Shipping Condition | Room Temperature |
Reactive Moiety | Dichlorotriazine |
Label or Dye | Texas Red™ |
Label Type | Classic Dyes |
Product Line | Texas Red™ |
Emission | 601 |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Molecular Probes™ Texas Red™ C2-Dichlorotriazine >
Spot an opportunity for improvement?Share a Content Correction