missing translation for 'onlineSavingsMsg'
Learn More

TFEC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13547-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16881814

  • 1355.00 SEK / 20µL
Estimated Shipment: 27-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the micropthalmia (MiT) family of basic helix-loop-helix leucine zipper transcription factors. MiT transcription factors regulate the expression of target genes by binding to E-box recognition sequences as homo- or heterodimers, and play roles in multiple cellular processes including survival, growth and differentiation. The encoded protein is a transcriptional activator of the nonmuscle myosin II heavy chain-A gene, and may also co-regulate target genes in osteoclasts as a heterodimer with micropthalmia-associated transcription factor. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications

Specifications

TFEC
Polyclonal
Unconjugated
TFEC
bHLHe34, hTFEC L, TCFEC, TFE C, TFEC, TFECL, transcription factor EC, Transcription factor EC like
Rabbit
Antigen Affinity Chromatography
RUO
21426, 22797, 26296
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14948, Q63302, Q9WTW4
Tfec
TFEC Fusion Protein Ag4061
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers