Learn More
TFG Mouse anti-Human, Unconjugated, Clone: 1B5B9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66916-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16889205
  • 1355.00 SEK / 20µL
Estimated Shipment: 30-03-2023
to see stock.
Description
Description
The TFG antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TFG. This antibody recognizes human, pig antigen. The TFG antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
TFG
Monoclonal
1000 μg/mL
Mouse
Primary
Pig, Human
ELISA, Immunohistochemistry, Western Blot
1B5B9
Unconjugated
20 μL
10342
IgG2b