Learn More
TFG Mouse anti-Human, Unconjugated, Clone: 1B5B9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66916-1-IG-20UL
Description
The TFG antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TFG. This antibody recognizes human, pig antigen. The TFG antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
TFG | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Pig |
Western Blot, Immunohistochemistry, ELISA | |
1B5B9 | |
Unconjugated | |
20 μL | |
10342 | |
IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.