missing translation for 'onlineSavingsMsg'
Learn More

TFII I Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen TFII I
Concentration 0.24 mg/mL
Applications Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16896913
View Documents
Proteintech
10499-1-AP-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16886913
View Documents
Proteintech
10499-1-AP-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11. 23. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7, 13 and 21.
Specifications

Specifications

TFII I
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunocytochemistry
Unconjugated
Rabbit
Rat, Human, Mouse
P78347, Q5U2Y1, Q9ESZ8
14886, 2969, 353256
TFII I Fusion Protein Ag0773
Primary
-20°C
GTF2I
0.24 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
BAP 135, BAP135, BTK associated protein 135, BTKAP1, DIWS, GTF2I, GTFII I, IB291, SPIN, SRF Phox1 interacting protein, TFII I, WBS, WBSCR6
Gtf2i
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
TFII I Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.