Learn More
Proteintech TFPI Mouse anti-Human, Unconjugated, Clone: 1B12F11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66842-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16889085
  • 1355.00 SEK / 20µL
Estimated Shipment: 05-06-2023
to see stock.
Description
Description
The TFPI antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TFPI. This antibody recognizes human antigen. The TFPI antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TFPI
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1B12F11
Unconjugated
20 μL
7035
IgG2a