missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFPT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | TFPT |
---|---|
Concentration | 0.42 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16886253
|
Proteintech
10097-2-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16876253
|
Proteintech
10097-2-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TFPT component of the chromatin-remodeling INO80 complex which, in addition to chromatin remodeling implicated in crucial functions such as DNA repair, checkpoint regulation, DNA replication, telomemere maintece and chromosome segregation. This protein appears to promote apoptosis in a TP53/p53-independent manner.Specifications
TFPT | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
P0C1Z6, Q3U1J1, Q9JMG6 | |
29844, 69714, 85423 | |
TFPT Fusion Protein Ag0131 | |
Primary | |
-20°C | |
TFPT |
0.42 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
amida, FB1, INO80 complex subunit F, INO80F, Protein FB1, TCF3 fusion partner, TFPT | |
Tfpt | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TFPT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech