missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGFBR2 Mouse anti-Human, Mouse, Rat, Clone: 2D5H7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66636-1-IG-150UL
This item is not returnable.
View return policy
Description
This gene encodes a subunit of mitochondrial ATP synthase. Mitochondrial ATP synthase catalyzes ATP synthesis, utilizing an electrochemical gradient of protons across the inner membrane during oxidative phosphorylation. ATP synthase is composed of two linked multi-subunit complexes: the soluble catalytic core, F1, and the membrane-spanning component, Fo, comprising the proton channel. The catalytic portion of mitochondrial ATP synthase consists of 5 different subunits (alpha, beta, gamma, delta, and epsilon) assembled with a stoichiometry of 3 alpha, 3 beta, and a single representative of the other 3. The proton channel consists of three main subunits (a, b, c). This gene encodes the gamma subunit of the catalytic core. Alternatively spliced transcript variants encoding different isoforms have been identified. This gene also has a pseudogene on chromosome 14.Specifications
TGFBR2 | |
Monoclonal | |
0.7 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P37173, P38438, Q62312 | |
TGFBR2 | |
TGFBR2 Fusion Protein Ag25773 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot | |
2D5H7 | |
Unconjugated | |
TGFBR2 | |
AAT3, FAA3, LDS1B, LDS2B, MFS2, RIIC, TAAD2, TbetaR II, TGF beta receptor type 2, TGF beta receptor type II, TGF beta type II receptor, TGFbeta RII, TGFBR2, TGFR 2 | |
Mouse | |
Protein G | |
RUO | |
21813, 7048, 81810 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TGFBR2 Mouse anti-Human, Mouse, Rat, Clone: 2D5H7, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction