missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGIF2LX Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17405-1-AP-20UL
This item is not returnable.
View return policy
Description
TGIF2LX is a member of the TALE/TGIF homeobox family of transcription factors. Testis-specific expression suggests that this gene may play a role in spermatogenesis. This gene encodes a member of the TALE/TGIF homeobox family of transcription factors. Testis-specific expression suggests that this gene may play a role in spermatogenesis. A homolog of this gene lies within the male specific region of chromosome Y, in a block of sequence that is thought to be the result of a large X-to-Y transposition.Specifications
TGIF2LX | |
Polyclonal | |
Unconjugated | |
TGIF2LX | |
Homeobox protein TGIF2LX, TGIF like on the X, TGIF2LX, TGIFLX | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
90316 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IUE1 | |
TGIF2LX | |
TGIF2LX Fusion Protein Ag11424 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TGIF2LX Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction